Learn More
Khk Enterprises Inc Aluminum Weighing Pans, 300 per Pack
Catalog No. NC9733999
Encompass
A pure aluminum sample vessel and weighing dish that is ideal as a Sample carrier for weighing equipment or as a container in MicroBalance applications where light weight and material purity are a must. Product dimension are 22x7x7mm (.0015 thick)
Catalog No. NC9733999 Supplier Khk Enterprises Inc Supplier No. C0290412/3
A pure aluminum sample vessel and weighing dish that is ideal as a Sample carrier for weighing equipment or as a container in MicroBalance applications where light weight and material purity are a must. Product dimension are 22x7x7mm (.0015 thick)