Learn More
Electron Microscopy Sciences Aluminum Wire, 0.25 mm, Purity 99.99% 10 FT
SDP
Catalog No. 5028060
Encompass_Preferred
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.25 mm
Purity: 99.99%
Catalog No. 50-280-60 Supplier Electron Microscopy Sciences Supplier No. 73000
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.25 mm
Purity: 99.99%