Learn More
USA Scientific Inc Aluminum single tab foil, standard size
Catalog No. NC9516817
Encompass
Aluminum single tab foil, pierceable, -80C to +120C, non-sterile, 100/box
Catalog No. NC9516817 Supplier USA Scientific Inc Supplier No. 29384100
Aluminum single tab foil, pierceable, -80C to +120C, non-sterile, 100/box