Learn More
MIRION TECHNOLOGIES CAPINTEC INC Tec-Control Aluminum Breakthru Kit
Catalog No. NC9470856
Encompass
.This item has a minimum order quantity of 6 per supplier requirements.
The Aluminum Break­thru Kit provides a ra­pid, easy and inexpensive way to test aqueous solutions, particularly per­tech­netate generator eluate, for trace quantities of alu­minum. Aluminum forms an intense red precipitate with the indicator paper, and the intensity of the color is directly proportional to the amount of aluminum in the solution. The USP allows a concentration of aluminum ion in an injection ≤ 10 micrograms per milliliter (10 µg/ml) in technetium 99m eluate prepared from Molybdenum 99 formed as a result of uranium fission.
Catalog No. NC9470856 Supplier MIRION TECHNOLOGIES CAPINTEC INC Supplier No. 150780
.This item has a minimum order quantity of 6 per supplier requirements.
The Aluminum Break­thru Kit provides a ra­pid, easy and inexpensive way to test aqueous solutions, particularly per­tech­netate generator eluate, for trace quantities of alu­minum. Aluminum forms an intense red precipitate with the indicator paper, and the intensity of the color is directly proportional to the amount of aluminum in the solution. The USP allows a concentration of aluminum ion in an injection ≤ 10 micrograms per milliliter (10 µg/ml) in technetium 99m eluate prepared from Molybdenum 99 formed as a result of uranium fission.