Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aluora™ Spatial Amplification Multiplex Dye Kit

Catalog No. ASHRP8PLEX
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 x 3-plex Slides
100 x 4-plex Slides
100 x 5-plex Slides
100 x 6-plex Slides
100 x 8-plex Slides
Conjugate:
Aluora 430, 488, 514, 555, 594, 647, 700, 750 Dyes
Aluora 488, 555, 594, 647, 700, 750 Dyes
Aluora 488, 555, 594, 647, 750 Dyes
Aluora 488, 555, 647 Dyes
Aluora 488, 555, 647, 750 Dyes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ASHRP8PLEX 100 x 8-plex Slides Aluora 430, 488, 514, 555, 594, 647, 700, 750 Dyes
ASHRP3PLEX 100 x 3-plex Slides Aluora 488, 555, 647 Dyes
ASHRP4PLEX 100 x 4-plex Slides Aluora 488, 555, 647, 750 Dyes
ASHRP5PLEX 100 x 5-plex Slides Aluora 488, 555, 594, 647, 750 Dyes
ASHRP6PLEX 100 x 6-plex Slides Aluora 488, 555, 594, 647, 700, 750 Dyes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ASHRP8PLEX Supplier Invitrogen™ Supplier No. ASHRP8PLEX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluora Spatial Amplification multiplex kits are optimized dye-only kits configured with 3 to 8 dyes for IHC, mIHC, and ISH panels.

Aluora™ Spatial Amplification multiplex kits are dye-only kits configured with 3 to 8 dyes providing a highly sensitive method for multiplex immunohistochemical (mIHC) and in-situ hybridization (ISH) analysis for use with the Aluora HRP reagents. The kits combine Aluora dye configurations for 3 to 8 multiplex panels compatible with fluorescence microscopy instruments and spectral imagers.

Revolutionary signal amplification technology

The Aluora spatial amplification kits represent an advance in signal amplification for spatial proteomics and transcriptomics. They employ proprietary Aluora reagents that react with horseradish peroxidase (HRP) to deposit photostable Aluora dyes onto target proteins, significantly improving the signal-to-background and helping reduce the impact of tissue autofluorescence in commonly imaged wavelengths. This chemistry enables robust multiplexed marker detection even in challenging tissue samples. Aluora kits requires 10–5000X less primary antibody than standard and multiplex IHC with antibodies alone and achieve greater or equal signal intensity.

Aluora spatial amplification reagents are optimized for use with Prolong Glass Antifade mountants and 1 mg/mL Thermo Scientific DAPI nucleic acid stains for multiplex analysis.

Aluora reagents and dyes can be purchased individually for use in IHC and ISH workflows or in pre-configured kits to detect mouse, rabbit, or biotin-labelled antibodies. The Aluora multiplex kits come as dye-only kits configured with 3 to 8 dyes or as the Aluora rainbow kit is configured with 8 Aluora dyes and 4X GAM and 4X GAR secondary detection core reagents for 100 slides in 8-plex.

Features of Aluora spatial amplification reagents include:

  • Signal amplification for enhanced detection sensitivity
  • Comprehensive spatial analysis and imaging capabilities for research applications
  • Fluorescent detection system for multiplexing and quantitative analysis
  • Flexible workflow options that accommodate diverse experimental needs
  • Polymerase HRP (PolyHRP) conjugated secondary antibodies that boost signal for low abundance targets
  • Highly cross-adsorbed secondary antibodies that help ensure target-specific detection and background reduction
  • Enhanced background reduction through proprietary blocking reagents for crisp, clear imaging

Optimized for automated slide staining

A major benefit of Aluora spatial amplification kits is their compatibility with automated slide stainers right out of the box. This enables complete labeling of a tissue slide with an 8-plex Aluora panel and DAPI with walk-away automation sample processing.

Reducing background for cleaner images

Aluora spatial amplification kits include blockers to help reduce endogenous peroxidase activity and autofluorescence. By addressing autofluorescence and background noise, less primary antibody is needed to generate a bright specific signal.

Iterative labeling enables unlimited multiplexing

The covalently bound Aluora dyes are resistant to antibody stripping, remaining brightly fluorescent through subsequent labeling cycles. This enables iterative staining of numerous targets using primary antibodies from the same host species—a major constraint in traditional immunohistochemistry. Any number of directly conjugated primary antibodies can also be introduced after Aluora labeling is complete.

Custom optimization for diverse applications

Although general protocols are provided, each sample requires optimization for optimal results. Tissue preparation and processing methods vary widely and can profoundly impact staining. Careful optimization of the complete workflow is critical to achieve the highest quality multiplexed data from precious samples. Aluora spatial amplification provides a versatile tool to address diverse spatial proteomics applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Dye Multiplex Kit
Clonality None
Target Species None
Quantity 100 x 8-plex Slides
For Use With (Application) Spatial Proteomics, Immunohistochemistry (IHC), IHC Staining and Detection, Fluorescent Staining, Immunofluorescence (IF)
Product Line Aluora
Conjugate Aluora 430, 488, 514, 555, 594, 647, 700, 750 Dyes
Detection Method Fluorescence
Form Lyophilized dye in screw-capped vials
Sample Type FFPE Tissue
Content And Storage 8 Aluora Dye Vials
•1 x Aluora 430, 1 x Aluora 488, 1 x Aluora 514, 1 x Aluora 555, 1 x Aluora 594, 1 x Aluora 647, 1 x Aluora 700, 1 x Aluora 750
•-25°C to -5°C
•Desiccate and protect from light.
Packaging Quantity 8 Pk.
Sufficient For Spatial Proteomics and Transcriptomics
Excitation/Emission 427/499, 493/518, 512/529, 553/567, 589/615, 652/670, 687/706, 757/783
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.