Learn More
Med Vet International Alushield Aerosol Bandage Spray 75 Gm
Catalog No. 502828818
Encompass_Preferred
AluShield Aerosol Bandage Spray is a convenient, water-resistant aerosol bandage that creates a protective barrier against external irritants in simple wounds of small and large animals. No antibiotic residues Allows the wound to breath Reduces risk of infection Active ingredient: Aluminum Powder 40 mg/g
Catalog No. 50-282-8818 Supplier Med Vet International Supplier No. PHVALUSHIELD
AluShield Aerosol Bandage Spray is a convenient, water-resistant aerosol bandage that creates a protective barrier against external irritants in simple wounds of small and large animals. No antibiotic residues Allows the wound to breath Reduces risk of infection Active ingredient: Aluminum Powder 40 mg/g