Learn More
Hach Company AluVer 3 Aluminum Reagent Powder Pillows, pk/100
Supplier:  Hach Company 1429099
Encompass
For aluminum determination by the aluminon method. Hach Method 8012. Range: up to 0.80 mg/L Al. Pack of 100 powder pillows.
Catalog No. 50-199-4987