Learn More
IDEXX LPD IDEXX ALV Ag Test, ELISA Ag, Antigen-capture, ALV p27 Antigen, Avian Leukosis Virus, Light albumin, cloacal swabs, serum, 5 plates/solid, Worldwide availability
Supplier:  IDEXX LPD 9909254
Encompass
The IDEXX ALV Ag Test is a highly specific antigen-capture ELISA designed for detecting avian leukemia virus (ALV) by targeting the p27 antigen, which is common across all subgroups. This reliable method accommodates various sample types, including serum and cloacal swabs, making it versatile for diagnostic applications in poultry health management.
  • Highly specific detection of avian leukemia virus
  • Robust antigen-capture ELISA assay format
  • Supports multiple sample types: Light albumin, cloacal swabs, serum
  • Packaging includes 5 plates per solid
  • Available worldwide for broad accessibility
Catalog No. NC9890603