Learn More
Apexbio Technology LLC Alvelestat 848141-11-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000091037
Encompass_Preferred
Alvelestat (CAS 848141-11-7) is an orally bioavailable reversible inhibitor of neutrophil elastase (NE) It demonstrates high affinity for human NE exhibiting a Ki of 9 4 nM and over 600-fold selectivity compared to other serine proteases In whole blood cell-based and degranulation assays alvelestat shows pIC50 values of 7 36 7 32 and 7 3 respectively Preclinical studies indicate that alvelestat reduces NE-induced markers of lung injury such as hemoglobin and hydroxyproline levels and attenuates neutrophilic inflammation in models of acute lung injury as well as smoke-induced airway inflammation and emphysema These properties support its use in research on neutrophil-driven pulmonary diseases
Catalog No. 50-000-91037 Supplier Apexbio Technology LLC Supplier No. B10375.1
Alvelestat (CAS 848141-11-7) is an orally bioavailable reversible inhibitor of neutrophil elastase (NE) It demonstrates high affinity for human NE exhibiting a Ki of 9 4 nM and over 600-fold selectivity compared to other serine proteases In whole blood cell-based and degranulation assays alvelestat shows pIC50 values of 7 36 7 32 and 7 3 respectively Preclinical studies indicate that alvelestat reduces NE-induced markers of lung injury such as hemoglobin and hydroxyproline levels and attenuates neutrophilic inflammation in models of acute lung injury as well as smoke-induced airway inflammation and emphysema These properties support its use in research on neutrophil-driven pulmonary diseases