Learn More
Selleck Chemical LLC Alvelestat (AZD9668) 25mg 848141-11-7 Avelestat
SDP
Catalog No. 501364533
Encompass_Preferred
Alvelestat (AZD9668, Avelestat) is an oral, highly selective inhibitor of neutrophil elastase (NE) with IC50 and Ki of 12 nM and 9.4 nM, at least 600-fold more selective over other serine proteases. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4533 Supplier Selleck Chemical LLC Supplier No. S721825MG
Alvelestat (AZD9668, Avelestat) is an oral, highly selective inhibitor of neutrophil elastase (NE) with IC50 and Ki of 12 nM and 9.4 nM, at least 600-fold more selective over other serine proteases. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.