Learn More
Selleck Chemical LLC Alvelestat (AZD9668) 5mg 848141-11-7 Avelestat
SDP
Catalog No. 501364532
Encompass_Preferred
Alvelestat (AZD9668, Avelestat) is an oral, highly selective inhibitor of neutrophil elastase (NE) with IC50 and Ki of 12 nM and 9.4 nM, at least 600-fold more selective over other serine proteases. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4532 Supplier Selleck Chemical LLC Supplier No. S72185MG
Add to Cart
Add to Cart
Alvelestat (AZD9668, Avelestat) is an oral, highly selective inhibitor of neutrophil elastase (NE) with IC50 and Ki of 12 nM and 9.4 nM, at least 600-fold more selective over other serine proteases. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.