Learn More
Cayman Chemical Alverine (citrate), Purity: ≥98%, CAS: 5560-59-8, 5 g
SDP
Catalog No. 502933656
Encompass_Preferred
An antispasmodic agent; suppresses contractions induced by potassium or ACh in isolated guinea pig detrusor smooth muscle; blocks spontaneous and vagal-stimulated colonic motility in rabbits; reduces the number of abdominal contractions induced by 5-HTP or high volume rectal distension in rats at 75 μg/animal
Catalog No. 50-293-3656 Supplier Cayman Chemical Supplier No. 268925 G
An antispasmodic agent; suppresses contractions induced by potassium or ACh in isolated guinea pig detrusor smooth muscle; blocks spontaneous and vagal-stimulated colonic motility in rabbits; reduces the number of abdominal contractions induced by 5-HTP or high volume rectal distension in rats at 75 μg/animal