Learn More
Medchemexpress LLC HY-B0500 500mg Medchemexpress, Alverine (citrate) CAS:5560-59-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0500500MG
Encompass_Preferred
Medchemexpress, HY-B0500 500mg Alverine (citrate) CAS:5560-59-8 Alverine citrate is a 5-HT1A receptor antagonist, with an IC50 of 101 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2566