Learn More
Medchemexpress LLC Alverine citrate | 5560-59-8 | 99.49% | 5 G
SDP
Catalog No. 5000408031
Encompass_Preferred
Alverine citrate is a 5-HT1A receptor antagonist with an IC50 of 101 nM. It is a drug used for functional gastrointestinal disorders and acts directly on the muscle in the gut, causing it to relax. It suppresses the effect of 5-HTP, but not that of 8-OH-DPAT. This compound is suitable for various laboratory applications.
  • Acts as a 5-HT1A receptor antagonist
  • Used for functional gastrointestinal disorders
  • Relaxes gut muscles directly
  • Suppresses the effect of 5-HTP
  • Has a high affinity for 5-HT1A receptors
Catalog No. 50-004-08031 Supplier Medchemexpress LLC Supplier No. HYB05005G
Alverine citrate is a 5-HT1A receptor antagonist with an IC50 of 101 nM. It is a drug used for functional gastrointestinal disorders and acts directly on the muscle in the gut, causing it to relax. It suppresses the effect of 5-HTP, but not that of 8-OH-DPAT. This compound is suitable for various laboratory applications.
  • Acts as a 5-HT1A receptor antagonist
  • Used for functional gastrointestinal disorders
  • Relaxes gut muscles directly
  • Suppresses the effect of 5-HTP
  • Has a high affinity for 5-HT1A receptors