Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alverine citrate | 5560-59-8 | 99.5% | 10 MM 1 ML
SDP

Catalog No. 5000406420
Encompass_Preferred

Alverine citrate is a 5-HT1A receptor antagonist with an IC50 of 101 nM, primarily used for research and analytical applications. It acts directly on gut muscles, causing relaxation, and is used in studies of functional gastrointestinal disorders. This compound shows high affinity for 5-HT1A receptors and weak affinity for 5-HT3 and 5-HT4 subtypes.

  • 5-HT1A receptor antagonist
  • IC50 of 101 nM
  • Used for research and analytical applications
  • Relaxes gut muscles
  • High affinity for 5-HT1A receptors
  • Molecular formula: C26H35NO7
  • Molecular weight: 473.56
  • Appearance: White to off-white solid

Catalog No. 50-004-06420 Supplier Medchemexpress LLC Supplier No. HYB050010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alverine citrate is a 5-HT1A receptor antagonist with an IC50 of 101 nM, primarily used for research and analytical applications. It acts directly on gut muscles, causing relaxation, and is used in studies of functional gastrointestinal disorders. This compound shows high affinity for 5-HT1A receptors and weak affinity for 5-HT3 and 5-HT4 subtypes.

  • 5-HT1A receptor antagonist
  • IC50 of 101 nM
  • Used for research and analytical applications
  • Relaxes gut muscles
  • High affinity for 5-HT1A receptors
  • Molecular formula: C26H35NO7
  • Molecular weight: 473.56
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.