Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alverine-d5 citrate | 1215327-00-6 | 98.5% | 1 MG
SDP

Catalog No. 5000409287
Encompass_Preferred

Alverine-d5 citrate is a deuterium-labeled analog of Alverine citrate, categorized as an isotope-labeled compound. It is a white to off-white solid with a purity of 98.5%. This compound can be used in research as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.

  • White to off-white solid appearance
  • Purity of 98.5%
  • Utilized as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS techniques
  • Molecular weight: 478.59
  • Chemical formula: C26H30D5NO7
  • Isotopic enrichment: 99.6%
  • Storage as powder: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C

Catalog No. 50-004-09287 Supplier Medchemexpress LLC Supplier No. HYB0500AS1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alverine-d5 citrate is a deuterium-labeled analog of Alverine citrate, categorized as an isotope-labeled compound. It is a white to off-white solid with a purity of 98.5%. This compound can be used in research as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.

  • White to off-white solid appearance
  • Purity of 98.5%
  • Utilized as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS techniques
  • Molecular weight: 478.59
  • Chemical formula: C26H30D5NO7
  • Isotopic enrichment: 99.6%
  • Storage as powder: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.