Learn More
Medchemexpress LLC Alverine-d5 citrate | 1215327-00-6 | 98.5% | 5 MG
SDP
Catalog No. 5000407994
Encompass_Preferred
Alverine-d5 (citrate) is the deuterium labeled Alverine citrate. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
Catalog No. 50-004-07994 Supplier Medchemexpress LLC Supplier No. HYB0500AS5MG
Alverine-d5 (citrate) is the deuterium labeled Alverine citrate. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development