Learn More
Abcam Alvespimycin (17-DMAG) hydrochloride, 25MG
Catalog No. 502682933
Encompass_Preferred
MW 653.2 Da, Purity >99%. A potent Hsp90 inhibitor (IC₅₀ = 62 nM), can bind to the ATP-binding motif and inhibit the protein chaperoning activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2933 Supplier Abcam Supplier No. AB27353125MG
MW 653.2 Da, Purity >99%. A potent Hsp90 inhibitor (IC₅₀ = 62 nM), can bind to the ATP-binding motif and inhibit the protein chaperoning activity.
The product is subject to the following: Abcam Restricted Use Statement