Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alvespimycin (17-DMAG) trifluoroacetate | 00-00-0 | MFCD08457919 | 99.0% | 730.77 g/mol | C34H49F3N4O10 | 5 MG
SDP

Catalog No. 5000243310
Encompass_Preferred

Alvespimycin TFA is the trifluoroacetate salt of 17-DMAG, a small-molecule inhibitor of heat shock protein 90 (Hsp90) used as a research reagent to study Hsp90-dependent signaling and anticancer mechanisms. The compound binds Hsp90 with an EC50 of 62 nM and is supplied as a high-purity, pale purple solid for laboratory use.

  • Potent Hsp90 inhibitor (EC50 62 nM).
  • Trifluoroacetate (TFA) salt form for consistent handling.
  • High purity (reported ~98.97%).
  • Molecular weight 730.77 g/mol.
  • Pale purple solid appearance.
  • Storage: sealed, away from moisture and light; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-43310 Supplier Medchemexpress LLC Supplier No. HY10389A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alvespimycin TFA is the trifluoroacetate salt of 17-DMAG, a small-molecule inhibitor of heat shock protein 90 (Hsp90) used as a research reagent to study Hsp90-dependent signaling and anticancer mechanisms. The compound binds Hsp90 with an EC50 of 62 nM and is supplied as a high-purity, pale purple solid for laboratory use.

  • Potent Hsp90 inhibitor (EC50 62 nM).
  • Trifluoroacetate (TFA) salt form for consistent handling.
  • High purity (reported ~98.97%).
  • Molecular weight 730.77 g/mol.
  • Pale purple solid appearance.
  • Storage: sealed, away from moisture and light; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.