Learn More
Selleck Chemical LLC Alvespimycin (17-DMAG) HCl S1142-5mg
SDP
Catalog No. 503108388
Encompass_Preferred
Alvespimycin (17-DMAG NSC 707545 BMS 826476 KOS 1022) HCl is a potent HSP90 inhibitor with IC50 of 62 nM in a cell-free assay
Catalog No. 50-310-8388 Supplier Selleck Chemical LLC Supplier No. S11425MG
Alvespimycin (17-DMAG NSC 707545 BMS 826476 KOS 1022) HCl is a potent HSP90 inhibitor with IC50 of 62 nM in a cell-free assay