Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alvespimycin hydrochloride | 467214-21-7 | 98.8% | 100 MG
SDP

Catalog No. 5000234434
Encompass_Preferred

Alvespimycin hydrochloride is the hydrochloride salt of 17-DMAG, a potent Hsp90 inhibitor used in biochemical and cell-based research. It binds Hsp90 with nanomolar potency and is typically supplied as a solid or as a solution in DMSO for in vitro studies.

  • High purity suitable for research (98.8%).
  • Molecular weight 653.21 g/mol.
  • Molecular formula C32H49ClN4O8.
  • Reported Hsp90 EC50 = 62 ± 29 nM; induces Hsp70 and promotes Her2 degradation in cell assays.
  • Available as solid or 10 mM solution in DMSO.
  • Intended for in vitro and cell-based assays.

Catalog No. 50-002-34434 Supplier Medchemexpress LLC Supplier No. HY12024100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alvespimycin hydrochloride is the hydrochloride salt of 17-DMAG, a potent Hsp90 inhibitor used in biochemical and cell-based research. It binds Hsp90 with nanomolar potency and is typically supplied as a solid or as a solution in DMSO for in vitro studies.

  • High purity suitable for research (98.8%).
  • Molecular weight 653.21 g/mol.
  • Molecular formula C32H49ClN4O8.
  • Reported Hsp90 EC50 = 62 ± 29 nM; induces Hsp70 and promotes Her2 degradation in cell assays.
  • Available as solid or 10 mM solution in DMSO.
  • Intended for in vitro and cell-based assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.