Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alvespimycin hydrochloride | 467214-21-7 | 98.8% | 5 MG
SDP

Catalog No. 5000231716
Encompass_Preferred

Alvespimycin hydrochloride is a small-molecule Hsp90 inhibitor used for biochemical and cell-based research. It is supplied as the hydrochloride salt to improve solubility and has reported activity at low nanomolar concentrations.

  • Potent Hsp90 inhibitor; EC50 62 ± 29 nM.
  • Hydrochloride salt for enhanced solubility.
  • Molecular weight 653.21 g/mol; formula C32H49ClN4O8.
  • Available in small pack sizes suitable for research-scale use.
  • Purity reported as 98.8% (HPLC).
  • Suitable for biochemical and cell-based assays.

Catalog No. 50-002-31716 Supplier Medchemexpress LLC Supplier No. HY120245MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alvespimycin hydrochloride is a small-molecule Hsp90 inhibitor used for biochemical and cell-based research. It is supplied as the hydrochloride salt to improve solubility and has reported activity at low nanomolar concentrations.

  • Potent Hsp90 inhibitor; EC50 62 ± 29 nM.
  • Hydrochloride salt for enhanced solubility.
  • Molecular weight 653.21 g/mol; formula C32H49ClN4O8.
  • Available in small pack sizes suitable for research-scale use.
  • Purity reported as 98.8% (HPLC).
  • Suitable for biochemical and cell-based assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.