Learn More
Medchemexpress LLC Alvespimycin trifluoroacetate | 00-00-0 | MFCD08457919 | 99.0% | 730.77 g/mol | C34H49F3N4O10 | 1 MG
SDP
Catalog No. 5000244154
Encompass_Preferred
Alvespimycin TFA is the trifluoroacetate salt of 17-DMAG, a small-molecule Hsp90 inhibitor used in research to probe Hsp90 biology, induce client protein degradation, and trigger stress responses in cancer cell models.
  • Potent Hsp90 inhibition with low-nanomolar cellular activity.
  • Promotes degradation of Hsp90 client proteins, including Her2.
  • Induces Hsp70 expression and apoptosis in sensitive cancer cells.
  • High purity solid suitable for biochemical and cellular assays.
  • Available in small research quantities for dose-response studies.
  • Stable when stored sealed at recommended temperatures.
Catalog No. 50-002-44154 Supplier Medchemexpress LLC Supplier No. HY10389A1MG
Alvespimycin TFA is the trifluoroacetate salt of 17-DMAG, a small-molecule Hsp90 inhibitor used in research to probe Hsp90 biology, induce client protein degradation, and trigger stress responses in cancer cell models.
  • Potent Hsp90 inhibition with low-nanomolar cellular activity.
  • Promotes degradation of Hsp90 client proteins, including Her2.
  • Induces Hsp70 expression and apoptosis in sensitive cancer cells.
  • High purity solid suitable for biochemical and cellular assays.
  • Available in small research quantities for dose-response studies.
  • Stable when stored sealed at recommended temperatures.