Learn More
TARGETMOL CHEMICALS INC ALVIMOPAN 10MG
SDP
Catalog No. 502581455
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alvimopan (LY 246736) is a synthetic trans-3 4-dimethyl-4-(3-hydroxyphenyl) piperidine with peripherally selective opioid mu receptor antagonist activity. Alvimopan is a selective and competitive antagonist at mu-opioid receptors found in myenteric and submucosal neurons and the immune cells of the lamina propria in the human gut. Upon administration this agent binds to mu-opioid receptors in the gut thereby reversing opioid-related disturbances in gut motility. Alvimopan is approximately three to nine times more potent than naloxone. purity: 99%
Catalog No. 50-258-1455 Supplier TARGETMOL CHEMICALS INC Supplier No. T124210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alvimopan (LY 246736) is a synthetic trans-3 4-dimethyl-4-(3-hydroxyphenyl) piperidine with peripherally selective opioid mu receptor antagonist activity. Alvimopan is a selective and competitive antagonist at mu-opioid receptors found in myenteric and submucosal neurons and the immune cells of the lamina propria in the human gut. Upon administration this agent binds to mu-opioid receptors in the gut thereby reversing opioid-related disturbances in gut motility. Alvimopan is approximately three to nine times more potent than naloxone. purity: 99%