Learn More
Selleck Chemical LLC Alvimopan dihydrate
SDP
Catalog No. 5000003496
Encompass_Preferred
Alvimopan (LY-246736) is a potent relatively nonselective opioid antagonist with Ki values of 0 77 4 4 and 40 nM for the and opioid receptors respectively displaying 100-fold selectivity over other aminergic G-protein-coupled receptors
Catalog No. 50-000-03496 Supplier Selleck Chemical LLC Supplier No. S57015MG
Alvimopan (LY-246736) is a potent relatively nonselective opioid antagonist with Ki values of 0 77 4 4 and 40 nM for the and opioid receptors respectively displaying 100-fold selectivity over other aminergic G-protein-coupled receptors