Learn More
Medchemexpress LLC Alvimopan monohydrate | 1383577-62-5 | 99.2% | C25H34N2O5 | 2 MG
SDP
Catalog No. 502082282
Encompass_Preferred
Alvimopan monohydrate is a potent, selective, orally active, and reversible μ-opioid receptor antagonist with an IC50 of 1.7 nM. It demonstrates selectivity for the μ-opioid receptor (Ki=0.47 nM) over κ- and δ-opioid receptors. It is used for the research of postoperative ileus. The salt form usually boasts enhanced water solubility and stability.
  • Potent, selective, orally active, and reversible μ-opioid receptor antagonist.
  • IC50 of 1.7 nM for μ-opioid receptor.
  • Shows selectivity for μ-opioid receptor over κ- and δ-opioid receptors.
  • Used for research of postoperative ileus.
  • Salt form offers enhanced water solubility and stability.
Catalog No. 50-208-2282 Supplier Medchemexpress LLC Supplier No. HY766572MG
Alvimopan monohydrate is a potent, selective, orally active, and reversible μ-opioid receptor antagonist with an IC50 of 1.7 nM. It demonstrates selectivity for the μ-opioid receptor (Ki=0.47 nM) over κ- and δ-opioid receptors. It is used for the research of postoperative ileus. The salt form usually boasts enhanced water solubility and stability.
  • Potent, selective, orally active, and reversible μ-opioid receptor antagonist.
  • IC50 of 1.7 nM for μ-opioid receptor.
  • Shows selectivity for μ-opioid receptor over κ- and δ-opioid receptors.
  • Used for research of postoperative ileus.
  • Salt form offers enhanced water solubility and stability.