Learn More
Apexbio Technology LLC ALW-II-41-27, 10mg CAS# 1186206-79-0
SDP
Supplier:  Apexbio Technology LLC A316510
Encompass_Preferred
ALW-II-41-27 is a potent inhibitor of EPH family kinases, with an IC50 value of 11 nM to EPHA2 [1] [2].EPH family proteins are key regulators of both disease and normal development. EPH receptors are involved in many intracellular signaling pathways such as PI3K/AKT/mTOR, RAS/RAF/MAPK, FAK, SRC, ABL, and RHO/RAC/CDC42 [2].In H358 cells, treatment with ALW-II-41-27 at a concentration of 1 M within 15 minutes impaired the tyrosine phosphorylation of the EPHA2 receptor and continued to inhibit the tyrosine phosphorylation through 6 hours. ALW-II-41-27 also dose-dependently inhibited the EPHA2 phosphorylation induced by ligand. When the EPHA2 was depleted by RNAi in NSCLC cell lines, cells were much less sensitive to ALW-II-41-27. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8391