Learn More
Selleck Chemical LLC ALW II-41-27-5mg
SDP
Supplier:  Selleck Chemical LLC S65155MG
Encompass_Preferred
ALW II-41-27 (compound 7) is a potent inhibitor of Eph receptor tyrosine kinase with Kd of 12 nM for EphA2.
Catalog No. 50-238-9896