Learn More
TARGETMOL CHEMICALS INC ALW-II-49-7 5MG
SDP
Catalog No. 502579270
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALW-II-49-7 is a selective and potent inhibitor of EphB2 kinase with an EC50 value of 40 nM in cell. purity: 99%
Catalog No. 50-257-9270 Supplier TARGETMOL CHEMICALS INC Supplier No. T677365MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALW-II-49-7 is a selective and potent inhibitor of EphB2 kinase with an EC50 value of 40 nM in cell. purity: 99%