Learn More
New England Biolabs, Inc. AlwI – 2500 units
SDP
Supplier:  New England Biolabs, Inc. R0513L
Encompass
A restriction endonuclease that recognizes the sequence GGATCNNNN^N_.
Catalog No. 50-812-159
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product