Learn More
New England Biolabs, Inc. AlwI – 500 units
SDP
Catalog No. 50812160
Encompass
A restriction endonuclease that recognizes the sequence GGATCNNNN^N_.
Catalog No. 50-812-160 Supplier New England Biolabs, Inc. Supplier No. R0513S
A restriction endonuclease that recognizes the sequence GGATCNNNN^N_.