Learn More
New England Biolabs, Inc. AlwNI – 500 units
SDP
Catalog No. 50812162
Encompass
A restriction endonuclease that recognizes the sequence CAG_NNN^CTG.
Catalog No. 50-812-162 Supplier New England Biolabs, Inc. Supplier No. R0514S
A restriction endonuclease that recognizes the sequence CAG_NNN^CTG.