Learn More
Cayman Chemical ALX 5407hydrochlorIde 10mg
SDP
Catalog No. 502990604
Encompass_Preferred
An inhibitor of GlyT1 (IC50 = 3 nM); selective for GlyT1 over GlyT2 (IC50 = >10 µM), as well as over NMDA receptors and the inhibitory glycine receptor at 100 µM; increases prefrontal cortical levels of glycine in rats at 10 mg/kg; reverses MK-801-induced persistent latent inhibition in a mouse model of schizophrenia
Catalog No. 50-299-0604 Supplier Cayman Chemical Supplier No. 3062610 MG
An inhibitor of GlyT1 (IC50 = 3 nM); selective for GlyT1 over GlyT2 (IC50 = >10 µM), as well as over NMDA receptors and the inhibitory glycine receptor at 100 µM; increases prefrontal cortical levels of glycine in rats at 10 mg/kg; reverses MK-801-induced persistent latent inhibition in a mouse model of schizophrenia