Learn More
Aviva Systems Biology Corp Alx1 Antibody - N-terminal region (ARP42868_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Alx1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI314867, C130005I02, Cart1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-040