Learn More
Aviva Systems Biology Corp ALX3 Antibody - N-terminal region (ARP37852_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37852_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ALX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse ALX3. Alternate names: ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-56451