Learn More
Aviva Systems Biology Corp ALX4 Antibody - middle region (ARP33239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ALX4. Alternate names: FPP, KIAA1788, PFM, PFM1, PFM2, FND2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54930