Learn More
Aviva Systems Biology Corp ALX4 Antibody - N-terminal region (ARP32356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALX4. Alternate names: FND2ApplicationWB. Species reactivity: Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54541