Learn More
Aviva Systems Biology Corp Alx4 Antibody - middle region (ARP36820_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36820_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Alx4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Alx4. Alternate names: lstApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-517