Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Alexa Fluor 488-Labeled Human CD7 Protein, His Tag - 25 μg

Catalog No. NC3787186
Encompass_Preferred

ACROBiosystems Alexa Fluor 488-labeled human CD7 protein with a C-terminal His tag, supplied as a 25 μg lyophilized vial. This fluorescently labeled recombinant extracellular domain of human CD7 is produced in HEK293 cells and is suitable for flow cytometry, antibody binding assays, and assay development.

  • Expressed in HEK293 (human 293) cells.
  • Contains amino acids Ala 26-Pro 180.
  • Contains a C-terminal polyhistidine tag.
  • Conjugated to Alexa Fluor 488 (excitation 488 nm; emission 517 nm).
  • Lyophilized from PBS, pH 7.4, with trehalose as protectant.
  • Calculated molecular weight ~18.3 kDa.
  • Purity greater than 90% by SDS-PAGE.
  • Supplied as 25 μg; alternative 200 μg available.

Catalog No. NC3787186 Supplier ACROBiosystems Supplier No. CD7HA2H825UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACROBiosystems Alexa Fluor 488-labeled human CD7 protein with a C-terminal His tag, supplied as a 25 μg lyophilized vial. This fluorescently labeled recombinant extracellular domain of human CD7 is produced in HEK293 cells and is suitable for flow cytometry, antibody binding assays, and assay development.

  • Expressed in HEK293 (human 293) cells.
  • Contains amino acids Ala 26-Pro 180.
  • Contains a C-terminal polyhistidine tag.
  • Conjugated to Alexa Fluor 488 (excitation 488 nm; emission 517 nm).
  • Lyophilized from PBS, pH 7.4, with trehalose as protectant.
  • Calculated molecular weight ~18.3 kDa.
  • Purity greater than 90% by SDS-PAGE.
  • Supplied as 25 μg; alternative 200 μg available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.