Learn More
Cayman Chemical Allylthiourea, Purity: ≥95%, CAS: 109-57-9, 100 g
SDP
Catalog No. 502988227
Encompass_Preferred
A nitritation inhibitor; inhibits nitritation in Nitrobacteriae by chelating the copper in the ammonia monooxygenase active site from 8-80 µM; has been used in the study of micropollutant degradation kinetics in soil
Catalog No. 50-298-8227 Supplier Cayman Chemical Supplier No. 30282100 G
A nitritation inhibitor; inhibits nitritation in Nitrobacteriae by chelating the copper in the ammonia monooxygenase active site from 8-80 µM; has been used in the study of micropollutant degradation kinetics in soil