Learn More
Iwai North America ALyS705: Chemically defined (Animal-free) medium for the expansion and/or activation of human lymphocytes. Size: 1L
Supplier:  Iwai North America 120P10
Encompass_Preferred
ALyS705 without IL-2 is Chemically defined (Animal-free) medium suitable for activating and/or expanding T cells from human lymphocytes. Compared to ALyS™505N, ALyS™705 achieves an animal-free culture environment by utilizing a high molecular polymer instead of albumin, which in turn reduces the shear stress generated between cells. Resulting ALyS™705 ideal for cell therapy and has shown higher proliferation rates than ALyS™505N.1. Tomida, A. et al. Inhibition of MEK pathway enhances the antitumor efficacy of chimeric antigen receptor T cells against neuroblastoma. Cancer Sci 112, 4026–4036 (2021) DOI: 10.1111/cas.150742. Mie, K., Shimada, T., Akiyoshi, H., Hayashi, A. & Ohashi, F. Change in peripheral blood lymphocyte count in dogs following adoptive immunotherapy using lymphokine-activated T killer cells combined with palliative tumor resection. Veterinary Immunology and Immunopathology 177, 58–63 (2016) DOI: 10.1016/j.vetimm.2016.06.007
Catalog No. NC2733020