Learn More
Cayman Chemical AlytSIntrIfluoROCTAte sa 10mg
SDP
Catalog No. 502946363
Encompass_Preferred
A neuropeptide with diverse biological activities; induces hypertension with tachyphylaxis and increases blood glucose levels in dogs; reduces gastric acid secretion in rats; induces anorexigenic effects in neonatal chicks
Catalog No. 50-294-6363 Supplier Cayman Chemical Supplier No. 3442810 MG
A neuropeptide with diverse biological activities; induces hypertension with tachyphylaxis and increases blood glucose levels in dogs; reduces gastric acid secretion in rats; induces anorexigenic effects in neonatal chicks