Learn More
Alzet ALZAID Chemical Compatibility Test Kit for Use with ALZET Osmotic Pumps
SDP
Catalog No. NC0014090
Encompass
The ALZAID Chemical Compatibility Test Kit allows researchers to determine the compatibility of solvents and agents with ALZET Osmotic Pumps. It is comprised of polymer spheres identical in chemical makeup and processing to the material used in the reservoir of ALZET pumps. The ALZAID test kit can be used to determine the compatibility of the solvent alone or agent-solvent combinations by following the recommended procedures. Each kit contains materials for evaluating four solutions.
Catalog No. NC0014090 Supplier Alzet Supplier No. 0004750
Add to Cart
Add to Cart
The ALZAID Chemical Compatibility Test Kit allows researchers to determine the compatibility of solvents and agents with ALZET Osmotic Pumps. It is comprised of polymer spheres identical in chemical makeup and processing to the material used in the reservoir of ALZET pumps. The ALZAID test kit can be used to determine the compatibility of the solvent alone or agent-solvent combinations by following the recommended procedures. Each kit contains materials for evaluating four solutions.