Learn More
Alzet ALZETTM Brain Infusion Kit 1, Materials for Brain Infusion from any ALZET Osmotic Pump, 28 ga Cannulae Extend 3-5 mm Below Skull Surface
SDP
Catalog No. NC9261047
Encompass
ALZETTM Brain Infusion Kits are designed specifically for use with ALZET pumps for targeted delivery to the central nervous system, such as for infusion into the cerebral ventricles for wider distribution or direct microperfusion of discrete brain structures. ALZET Brain Infusion Kit 1 is optimized to reach the rat lateral ventricles or other structures of similar depth. The cannulae are a narrower design and extend 3-5 mm from the skull surface.Each ALZET Brain Infusion Kit 1 includes materials for 10 brain infusions:•10 Brain Infusion Cannulae (28 gauge, extend 3-5 mm from skull surface)•10 Vinyl Catheter Tubes•40 Depth-Adjustment Spacers•1 Instruction SheetKit is sterilized by gamma irradiation and ready for use.
Catalog No. NC9261047 Supplier Alzet Supplier No. 0004760
ALZETTM Brain Infusion Kits are designed specifically for use with ALZET pumps for targeted delivery to the central nervous system, such as for infusion into the cerebral ventricles for wider distribution or direct microperfusion of discrete brain structures. ALZET Brain Infusion Kit 1 is optimized to reach the rat lateral ventricles or other structures of similar depth. The cannulae are a narrower design and extend 3-5 mm from the skull surface.Each ALZET Brain Infusion Kit 1 includes materials for 10 brain infusions:•10 Brain Infusion Cannulae (28 gauge, extend 3-5 mm from skull surface)•10 Vinyl Catheter Tubes•40 Depth-Adjustment Spacers•1 Instruction SheetKit is sterilized by gamma irradiation and ready for use.