Learn More
Alzet ALZET Model 2001D Mini-Osmotic Pump, Continuous Infusion, Sterilized for Single Use, 200 µL
SDP
Catalog No. NC3312605
Encompass
The ALZET Model 2001D is a miniature, implantable osmotic pump designed to deliver solutions continuously in laboratory animals for research applications. Its self-powered design eliminates external connections, reducing animal stress and ensuring accurate dosing. The pump operates independently of drug formulation, features a nominal pumping rate of 8 µl/hr, and is intended for single use, making it ideal for precise experimental control in various anatomical sites.
Catalog No. NC3312605 Supplier Alzet Supplier No. 0000294
The ALZET Model 2001D is a miniature, implantable osmotic pump designed to deliver solutions continuously in laboratory animals for research applications. Its self-powered design eliminates external connections, reducing animal stress and ensuring accurate dosing. The pump operates independently of drug formulation, features a nominal pumping rate of 8 µl/hr, and is intended for single use, making it ideal for precise experimental control in various anatomical sites.