Learn More
Alzet ALZET Osmotic Pumps, Model 1004: 100 µl Volume, 0.11 µl/hr Release Rate, 28 Days Duration - for Orders of 10-40 Pumps
SDP
Catalog No. NC0836853
Encompass
ALZET Osmotic Pumps are small infusion devices for accurate, continuous administration of test agents in unrestrained lab animals. They offer a convenient alternative for long-term dosing compared to stress-inducing methods such as injections and gavage. Pumps are sold empty and easily filled with the test agent via syringe. Their pumping rate is determined at manufacture and is not affected by the molecular weight of the test agent. Pumps are implanted subcutaneously or intraperitoneally for systemic delivery, or can be attached to an ALZET catheter for targeted delivery such as to the brain, vasculature, etc. Sterilized by gamma irradiation and ready for use.Nominal Specifications for Model 1004:•Reservoir Capacity:100 µl•Release Rate:0.11 µl/hr•Duration:28 days•Minimum animal size:10 g mouseNote: Actual specifications vary within limits by lot and are provided with the pumps.See other listings for additional pump models including durations up to 6 weeks.
Catalog No. NC0836853 Supplier Alzet Supplier No. 0009922PK
ALZET Osmotic Pumps are small infusion devices for accurate, continuous administration of test agents in unrestrained lab animals. They offer a convenient alternative for long-term dosing compared to stress-inducing methods such as injections and gavage. Pumps are sold empty and easily filled with the test agent via syringe. Their pumping rate is determined at manufacture and is not affected by the molecular weight of the test agent. Pumps are implanted subcutaneously or intraperitoneally for systemic delivery, or can be attached to an ALZET catheter for targeted delivery such as to the brain, vasculature, etc. Sterilized by gamma irradiation and ready for use.Nominal Specifications for Model 1004:•Reservoir Capacity:100 µl•Release Rate:0.11 µl/hr•Duration:28 days•Minimum animal size:10 g mouseNote: Actual specifications vary within limits by lot and are provided with the pumps.See other listings for additional pump models including durations up to 6 weeks.