Learn More
Medchemexpress LLC HY-108329 5mg Medchemexpress, AM-0902 CAS:1883711-97-4 Purity:>98%
SDP
Catalog No. 501968530
Encompass_Preferred
Medchemexpress, HY-108329 5mg AM-0902 CAS:1883711-97-4 AM-0902 is a potent, selective transient receptor potential A1 (TRPA1) antagonist with IC50s of 71 and 131 nM for rTRPA1 and hTRPA1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8530 Supplier Medchemexpress LLC Supplier No. HY1083295MG
Medchemexpress, HY-108329 5mg AM-0902 CAS:1883711-97-4 AM-0902 is a potent, selective transient receptor potential A1 (TRPA1) antagonist with IC50s of 71 and 131 nM for rTRPA1 and hTRPA1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.