Learn More
LaMotte™ AM-12 TesTabs™ Water Investigation
SDPSCI_ED
Catalog No. S66982 Shop All LaMotte Company Products
Change view
Click to view available options
For Use With (Application):
Water Quality
Catalog No. For Use With (Application)
S66982 Water Quality
1 options
Catalog No. S66982 Supplier LaMotte™ Supplier No. 5849
Includes
Tablets to perform 100 tests per reagent module, test factor flashcards, color chart, test tubes and reference book (The Monitor's Handbook)
Test water quality as a complete experiment or in addition to other water studies.
  • Measures water quality with 12 TesTabs reagents
  • Uses easy-to-perform, nonhazardous TesTabs
  • Flashcards inform about each test factor, describe the chemical reaction and explain the chemical influence on the environment
  • To use: fill tube with water, add necessary tablets, shake tube until each tablet fully dissolves and compare resulting color to the color chart
Order Info
Grades 5+.
Specifications
For Use With (Application) Water Quality
Includes Test Modules for Alkalinity, Ammonia, Chloride, Chlorine, Chromium, Copper, Dissolved Oxygen/BOD, Hardness, Iron, Nitrate, pH, and Phosphate
WARNING: Cancer - www.P65Warnings.ca.gov