Learn More
TARGETMOL CHEMICALS INC AM-1235 5MG
SDP
Catalog No. 502580443
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AM-1235 is a potent and selective cannabinoid receptor CB1 agonist. purity: 98%
Catalog No. 50-258-0443 Supplier TARGETMOL CHEMICALS INC Supplier No. T299305MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AM-1235 is a potent and selective cannabinoid receptor CB1 agonist. purity: 98%