Learn More
Medchemexpress LLC HY-13467 5mg Medchemexpress, AM-1638 CAS:1142214-62-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134675MG
Encompass_Preferred
Medchemexpress, HY-13467 5mg AM-1638 CAS:1142214-62-7 AM-1638 is a potent and orally bioavailable GPR40/FFA1 full agonist with an EC50 of 0.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9855